1-Methyl-1-phenyl-2-thiourea 97%

Leverantör: Apollo Scientific

Synonyms: N-Methyl-N-phenylthiourea

OR9376-10G OR9376-50G
APOSOR9376-10GEA 2150 SEK
APOSOR9376-10G APOSOR9376-50G
1-Methyl-1-phenyl-2-thiourea 97%
1-Methyl-1-phenyl-2-thiourea
Formel: C₈H₁₀N₂S
MW: 166,24 g/mol
Smältpunkt: 100…104 °C
Storage Temperature: Omgivande
MDL Number: MFCD00086629
CAS nummer: 4104-75-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR