1,5,6,7-Tetrahydro-2-methyl-4H-indol-4-one 95%
Leverantör: Apollo Scientific
Synonyms: 2-Methyl-4-oxo-4,5,6,7-tetrahydroindole
OR937550-1G OR937550-25G OR937550-5G
APOSOR937550-1GEA 1475 SEK
APOSOR937550-1G APOSOR937550-25G APOSOR937550-5G
1,5,6,7-Tetrahydro-2-methyl-4H-indol-4-one 95%
1,5,6,7-Tetrahydro-2-methyl-4H-indol-4-one
Formel: C₉H₁₁NO
MW: 149.19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04972500
CAS nummer: 35308-68-0