5H-5-Methyl-6,7-dihydrocyclopentapyrazine 95%
Leverantör: Apollo Scientific
Synonyms: 5-Methyl-6,7-dihydro-5H-cyclopenta[b]pyrazine, 5-Methyl-2,3-cyclopentenopyrazine
OR937238-100G OR937238-10G OR937238-25G
APOSOR937238-100GEA 2325 SEK
APOSOR937238-100G APOSOR937238-10G APOSOR937238-25G
5H-5-Methyl-6,7-dihydrocyclopentapyrazine 95%
5H-5-Methyl-6,7-dihydrocyclopentapyrazine
Formel: C₈H₁₀N₂
MW: 134,18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00040997
CAS nummer: 23747-48-0