(S)-(-)-1-(2-Naphthyl)ethylamine 95%
Leverantör: Apollo Scientific
Synonyms: (S)-1-(Naphthalen-2-yl)ethanamine
OR936986-1G OR936986-5G
APOSOR936986-1GEA 1625 SEK
APOSOR936986-1G APOSOR936986-5G
(S)-(-)-1-(2-Naphthyl)ethylamine 95%
(S)-(-)-1-(2-Naphthyl)ethylamine
Formel: C₁₂H₁₃N
MW: 171,24 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00085366
CAS nummer: 3082-62-0