Azoxibenzen 98%

Leverantör: Apollo Scientific

OR936835-1G OR936835-25G OR936835-5G
APOSOR936835-1GEA 260 SEK
APOSOR936835-1G APOSOR936835-25G APOSOR936835-5G
Azoxibenzen 98%
Azoxibenzen
Formel: C₁₂H₁₀N₂O
Storage Temperature: Omgivande
MDL Number: MFCD00019925
CAS nummer: 495-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR