2-Methylbenzylamine 98%
Leverantör: Apollo Scientific
Synonyms: o-Tolylmethanamine, 2-(Aminomethyl)toluene
OR936728-100G OR936728-25G
APOSOR936728-100GEA 2200 SEK
APOSOR936728-100G APOSOR936728-25G
2-Methylbenzylamine 98%
2-Methylbenzylamine
Formel: C₈H₁₁N
MW: 121,18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00008112
CAS nummer: 89-93-0