4-Ethylphenyl isocyanate 95%
Leverantör: Apollo Scientific
Synonyms: 1-Ethyl-4-isocyanatobenzene
OR936429-25G OR936429-5G
APOSOR936429-25GEA 3500 SEK
APOSOR936429-25G APOSOR936429-5G
4-Ethylphenyl isocyanate 95%
4-Ethylphenyl isocyanate
  • May form small amount of sediment.
Formel: C₉H₉NO
MW: 147.18 g/mol
Densitet: 1.024
Storage Temperature: Omgivande
MDL Number: MFCD00013881
CAS nummer: 23138-50-3