5-Methoxyresorcinol 98%

Leverantör: Apollo Scientific

Synonyms: 5-Methoxybenzene-1,3-diol, 3,5-Dihydroxyanisole, Phloroglucinol Monomethyl Ether

OR936313-1G OR936313-5G OR936313-25G
APOSOR936313-1GEA 535 SEK
APOSOR936313-1G APOSOR936313-5G APOSOR936313-25G
5-Methoxyresorcinol 98%
5-Methoxyresorcinol
Formel: C₇H₈O₃
MW: 140,14 g/mol
Kokpunkt: 188…189 °C/12mmHg
Smältpunkt: 78…80 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002285
CAS nummer: 2174-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR