2-Propyl-1,3-propanediol 95%

Leverantör: Apollo Scientific

Synonyms: 2-Propylpropane-1,3-diol

OR936294-1G OR936294-5G
APOSOR936294-1GEA 805 SEK
APOSOR936294-1G APOSOR936294-5G
2-Propyl-1,3-propanediol 95%
2-Propyl-1,3-propanediol
Formel: C₆H₁₄O₂
MW: 118,18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00799394
CAS nummer: 2612-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR