cis-4-tert-Butylcyclohexanecarboxylic acid 95%
Leverantör: Apollo Scientific
OR936255-10G OR936255-1G OR936255-5G
APOSOR936255-10GEA 10425 SEK
APOSOR936255-10G APOSOR936255-1G APOSOR936255-5G
cis-4-tert-Butylcyclohexanecarboxylic acid 95%
cis-4-tert-Butylcyclohexanecarboxylic acid
Formel: C₁₁H₂₀O₂
Storage Temperature: Omgivande
CAS nummer: 943-28-2