1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers) 95%
Leverantör: Apollo Scientific
Synonyms: 1,4-Cyclohexanedimethanol
OR936250-100G OR936250-1KG OR936250-500G
APOSOR936250-100GEA 260 SEK
APOSOR936250-100G APOSOR936250-1KG APOSOR936250-500G
1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers) 95%
1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers)
Formel: C₈H₁₆O₂
Kokpunkt: 283 °C
Smältpunkt: 35 °C
Densitet: 1,083 g/cm³ (25 °C)
Flampunkt: 161 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001512
CAS nummer: 105-08-8