2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
Leverantör: Apollo Scientific
Varning
OR9352-1G OR9352-5G
APOSOR9352-1GEA 2075 SEK
APOSOR9352-1G APOSOR9352-5G
2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
Formel: C₁₀H₁₀N₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD00269632
CAS nummer: 874605-59-1