4-Formyltetrahydropyran 95%
Leverantör: Apollo Scientific
Varning
OR9347-1G OR9347-25G OR9347-5G OR9347-100G
APOSOR9347-1GEA 380 SEK
APOSOR9347-1G APOSOR9347-25G APOSOR9347-5G APOSOR9347-100G
4-Formyltetrahydropyran 95%
4-Formyltetrahydropyran
Formel: C₆H₁₀O₂
Storage Temperature: Kylskåp
MDL Number: MFCD03425264
CAS nummer: 50675-18-8