5-Norbornene-2,3-dimethanol 95%
Leverantör: Apollo Scientific
Synonyms: 2,3-Bis(hydroxymethyl)-5-norbornene, Bicyclo[2.2.1]hept-5-ene-2,3-dimethanol
OR933395-1G OR933395-5G
APOSOR933395-1GEA 890 SEK
APOSOR933395-1G APOSOR933395-5G
5-Norbornene-2,3-dimethanol 95%
5-Norbornene-2,3-dimethanol
Formel: C₉H₁₄O₂
MW: 154.21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00474435
CAS nummer: 85-39-2