3,3-Dimethylpiperidin-4-one hydrochloride 95%
Leverantör: Apollo Scientific
OR932950-1G OR932950-250MG
APOSOR932950-1GEA 1275 SEK
APOSOR932950-1G APOSOR932950-250MG
3,3-Dimethylpiperidin-4-one hydrochloride 95%
3,3-Dimethylpiperidin-4-one hydrochloride
Formel: C₇H₁₄ClNO
MW: 163.65 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11043097
CAS nummer: 648921-37-3