2,3-Dichloro-4-pyridineboronic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2,3-Dichloropyridine-4-boronic acid, 2,3-Dichloro-4-pyridylboronic acid
OR9329-1G OR9329-5G
APOSOR9329-1GEA 790 SEK
APOSOR9329-1G APOSOR9329-5G
2,3-Dichloro-4-pyridineboronic acid
2,3-Dichloro-4-pyridineboronic acid
Formel: C₅H₄BCl₂NO₂
MW: 191,81 g/mol
Smältpunkt: 136…138 °C
Storage Temperature: Omgivande
MDL Number: MFCD04038756
CAS nummer: 951677-39-7