1-Naphthylacetonitrile 95%
Leverantör: Apollo Scientific
OR932811-25G
OR932811-100G
APOSOR932811-25GEA
0
SEK
APOSOR932811-25G
APOSOR932811-100G
1-Naphthylacetonitrile 95%
1-Naphthylacetonitrile
Formel:
C₁₂H₉N Storage Temperature: Omgivande |
MDL Number:
MFCD00004041 CAS nummer: 132-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...