(4-Methoxybenzyl)methyl ether 98%
Leverantör: Apollo Scientific
Synonyms: 1-Methoxy-4-(methoxymethyl)benzene, 4-(Methoxymethyl)anisole
OR932402-25G OR932402-5G
APOSOR932402-25GEA 3950 SEK
APOSOR932402-25G APOSOR932402-5G
(4-Methoxybenzyl)methyl ether 98%
(4-Methoxybenzyl)methyl ether
Formel: C₉H₁₂O₂
MW: 152.19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00155135
CAS nummer: 1515-81-7