α,α'-Dichloro-p-xylene 95%
Leverantör: Apollo Scientific
Fara
Synonyms: 1,4-Bis(chloromethyl)benzene, p-Xylene dichloride, p-Xylylene dichloride
OR9311-100G OR9311-500G
APOSOR9311-100GEA 1950 SEK
APOSOR9311-100G APOSOR9311-500G
α,α'-Dichloro-p-xylene 95%
α,α'-Dichloro-p-xylene
Intermediate for organic synthesis
Formel: C₈H₈Cl₂
MW: 175,06 g/mol
Kokpunkt: 254 °C
Smältpunkt: 98…101 °C
Densitet: 1,417 g/cm³ (25 °C)
Flampunkt: 143 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000920
CAS nummer: 623-25-6
EINECS: 210-782-7
UN: 2928
ADR: 6.1+8,II