m-Tolyldiethanolamine 98%

Leverantör: Apollo Scientific

Synonyms: 2,2'-(3-Methylphenylimino)diethanol, 2,2'-(m-Tolylimino)diethanol, N,N-Bis(2-hydroxyethyl)-m-toluidine, 2,2'-(m-Tolylazanediyl)diethanol

OR930972-100G OR930972-500G
APOSOR930972-100GEA 1975 SEK
APOSOR930972-100G APOSOR930972-500G
m-Tolyldiethanolamine 98%
m-Tolyldiethanolamine
Formel: C₁₁H₁₇NO₂
MW: 195,26 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00020578
CAS nummer: 91-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR