2-Methoxy-3,5-dimethylbenzaldehyde 95%
Leverantör: Apollo Scientific
OR930885-1G OR930885-250MG
APOSOR930885-1GEA 8400 SEK
APOSOR930885-1G APOSOR930885-250MG
2-Methoxy-3,5-dimethylbenzaldehyde 95%
2-Methoxy-3,5-dimethylbenzaldehyde
Formel: C₁₀H₁₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD06247962
CAS nummer: 16313-77-2