2-tert-Butyl-4-methoxyphenol 95%
Leverantör: Apollo Scientific
Synonyms: 3-BHA, 4-Hydroxy-3-tert-butylanisole, 3-tert-Butyl-4-hydroxyanisole
OR930692-100G OR930692-25G OR930692-500G
APOSOR930692-100GEA 775 SEK
APOSOR930692-100G APOSOR930692-25G APOSOR930692-500G
2-tert-Butyl-4-methoxyphenol 95%
2-tert-Butyl-4-methoxyphenol
Formel: C₁₁H₁₆O₂
MW: 180,25 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00040484
CAS nummer: 121-00-6