5-Norbornene-2-methylamine (mixture of isomers) 95%
Leverantör: Apollo Scientific
Synonyms:
5-Norbornene-2-methylamine, 5-(Aminomethyl)bicyclo[2.2.1]hept-2-ene, 5-(Aminomethyl)norbornene, 5-Norbornene-2-methanamine
OR930316-1G
OR930316-250MG
APOSOR930316-1GEA
7625
SEK
APOSOR930316-1G
APOSOR930316-250MG
5-Norbornene-2-methylamine (mixture of isomers) 95%
5-Norbornene-2-methylamine (mixture of isomers)
Formel:
C₈H₁₃N MW: 123,20 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD07369043 CAS nummer: 95-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...