Methyl-3-chloro-1H-pyrrole-2-carboxylate 98%
Leverantör: Apollo Scientific
Synonyms: Methyl 3-chloro-1H-pyrrole-2-carboxylate
OR929899-10G OR929899-1G OR929899-5G
APOSOR929899-10GEA 3650 SEK
APOSOR929899-10G APOSOR929899-1G APOSOR929899-5G
Methyl-3-chloro-1H-pyrrole-2-carboxylate 98%
Methyl-3-chloro-1H-pyrrole-2-carboxylate
Formel: C₆H₆ClNO₂
MW: 159,57 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD20661619
CAS nummer: 226410-00-0