1-Methyl-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid 95%
Leverantör: Apollo Scientific
OR929858-1G OR929858-250MG
APOSOR929858-1GEA 5750 SEK
APOSOR929858-1G APOSOR929858-250MG
1-Methyl-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid 95%
1-Methyl-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid
Formel: C₈H₇N₃O₂
Storage Temperature: Omgivande
MDL Number: MFCD11109332
CAS nummer: 116855-09-5