Methyl-2-hydroxyisobutyrate 95%

Leverantör: Apollo Scientific

Synonyms: Methyl 2-Hydroxyisobutyrate, 2-Hydroxyisobutyric Acid Methyl Ester

OR929842-100G OR929842-25G
APOSOR929842-100GEA 2250 SEK
APOSOR929842-100G APOSOR929842-25G
Methyl-2-hydroxyisobutyrate 95%
Methyl-2-hydroxyisobutyrate
Formel: C₅H₁₀O₃
MW: 118,13 g/mol
Kokpunkt: 137 °C
Flampunkt: 41 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004457
CAS nummer: 2110-78-3
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR