N,N-Diethylcyclohexylamine 95%

Leverantör: Apollo Scientific

OR929820-100G OR929820-25G OR929820-500G
APOSOR929820-100GEA 2575 SEK
APOSOR929820-100G APOSOR929820-25G APOSOR929820-500G
N,N-Diethylcyclohexylamine 95%
N,N-Diethylcyclohexylamine
Formel: C₁₀H₂₁N
Storage Temperature: Omgivande
MDL Number: MFCD00003847
CAS nummer: 91-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR