3-Thiazol-2-ylbenzaldehyde 95%
Leverantör: Apollo Scientific
Synonyms:
3-Thiazol-2-yl-benzaldehyde
OR929751-1G
OR929751-250MG
APOSOR929751-1GEA
2750
SEK
APOSOR929751-1G
APOSOR929751-250MG
3-Thiazol-2-ylbenzaldehyde 95%
3-Thiazol-2-ylbenzaldehyde
Formel:
C₁₀H₇NOS MW: 189,24 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD06797781 CAS nummer: 885465-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...