2-Amino-1-pyrrolidine hydrochloride 98%

Leverantör: Apollo Scientific

Synonyms: 2-Amino-1-pyrroline hydrochloride, 3,4-Dihydro-2H-pyrrol-5-amine hydrochloride

OR929570-10G OR929570-1G OR929570-5G
APOSOR929570-10GEA 5475 SEK
APOSOR929570-10G APOSOR929570-1G APOSOR929570-5G
2-Amino-1-pyrrolidine hydrochloride 98%
2-Amino-1-pyrrolidine hydrochloride
Formel: C₄H₉ClN₂
MW: 120,58 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00790772
CAS nummer: 7544-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR