4-(4-Nitrophenyl)oxazole 97%
Leverantör: Apollo Scientific
OR929279-10G OR929279-1G OR929279-5G
APOSOR929279-10GEA 6775 SEK
APOSOR929279-10G APOSOR929279-1G APOSOR929279-5G
4-(4-Nitrophenyl)oxazole 97%
4-(4-Nitrophenyl)oxazole
Formel: C₉H₆N₂O₃
MW: 190,1600 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09032979
CAS nummer: 13382-61-1