Ac-Ile-OMe 95%

Leverantör: Apollo Scientific

OR929225-1G OR929225-5G
APOSOR929225-1GEA 810 SEK
APOSOR929225-1G APOSOR929225-5G
Ac-Ile-OMe 95%
Ac-Ile-OMe
Formel: C₉H₁₇NO₃
Storage Temperature: Omgivande
MDL Number: MFCD00080833
CAS nummer: 2256-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR