[3-(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)propyl]amine 95%
Leverantör: Apollo Scientific
OR927585-1G
OR927585-250MG
APOSOR927585-1GEA
8475
SEK
APOSOR927585-1G
APOSOR927585-250MG
[3-(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)propyl]amine 95%
[3-(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)propyl]amine
Formel:
C₈H₁₄ClN₃ MW: 187,67 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD03419761 CAS nummer: 956786-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...