Methyl-5-chloro-2-methylnicotinate 98%

Leverantör: Apollo Scientific

Synonyms: Methyl 5-chloro-2-methylnicotinate

OR926987-1G OR926987-250MG
APOSOR926987-1GEA 5475 SEK
APOSOR926987-1G APOSOR926987-250MG
Methyl-5-chloro-2-methylnicotinate 98%
Methyl-5-chloro-2-methylnicotinate
Formel: C₈H₈ClNO₂
MW: 185,6100 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09908061
CAS nummer: 350597-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR