(2E)-3-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)acrylic acid 95%

Leverantör: Apollo Scientific

Synonyms: 3-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)prop-2-enoic acid

OR926704-1G OR926704-250MG
APOSOR926704-1GEA 8475 SEK
APOSOR926704-1G APOSOR926704-250MG
(2E)-3-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)acrylic acid 95%
(2E)-3-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)acrylic acid
Formel: C₉H₁₂N₂O₂
MW: 180,21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02090856
CAS nummer: 512809-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR