N,N,3,5-Tetramethylaniline 95%
Leverantör: Apollo Scientific
Synonyms: N,N-Dimethyl-3,5-xylidine, 3,5,N,N-Tetramethylaniline, 3,5-Dimethyl-N,N-dimethylaniline
OR926672-100G OR926672-25G OR926672-5G
APOSOR926672-100GEA 8925 SEK
APOSOR926672-100G APOSOR926672-25G APOSOR926672-5G
N,N,3,5-Tetramethylaniline 95%
N,N,3,5-Tetramethylaniline
Formel: C₁₀H₁₅N
MW: 149,24 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00008307
CAS nummer: 4913-13-7