4-Chloro-5,7-dihydro-6H-pyrrolo[2,3-d]pyrimidin-6-one 96%
Leverantör: Apollo Scientific
Synonyms: 4-chloro-5H,6H,7H-pyrrolo[2,3-d]pyrimidin-6-one, 4-Chloro-5H-pyrrolo[2,3-d]pyrimidin-6(7H)-one
OR925978-1G OR925978-250MG OR925978-5G
APOSOR925978-1GEA 2575 SEK
APOSOR925978-1G APOSOR925978-250MG APOSOR925978-5G
4-Chloro-5,7-dihydro-6H-pyrrolo[2,3-d]pyrimidin-6-one 96%
4-Chloro-5,7-dihydro-6H-pyrrolo[2,3-d]pyrimidin-6-one
Formel: C₆H₄ClN₃O
MW: 169,57 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09832903
CAS nummer: 346599-63-1