Ethyl (3-Hydroxyphenyl)acetate 95%

Leverantör: Apollo Scientific

Synonyms: Ethyl 2-(3-hydroxyphenyl)acetate

OR925825-1G OR925825-250MG
APOSOR925825-1GEA 635 SEK
APOSOR925825-1G APOSOR925825-250MG
Ethyl (3-Hydroxyphenyl)acetate 95%
Ethyl (3-Hydroxyphenyl)acetate
Formel: C₁₀H₁₂O₃
MW: 180,20 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07369387
CAS nummer: 22446-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR