2-Methyl-2-propyl-1,3-propanediol 95%
Leverantör: Apollo Scientific
Synonyms:
2,2-Bis(hydroxymethyl)pentane, 2-Methyl-2-propylpropane-1,3-diol, 2,2-Dimethylolpentane
OR925815-25G
OR925815-100G
APOSOR925815-25GEA
1475
SEK
APOSOR925815-25G
APOSOR925815-100G
2-Methyl-2-propyl-1,3-propanediol 95%
2-Methyl-2-propyl-1,3-propanediol
Formel:
C₇H₁₆O₂ MW: 132,2 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00004696 CAS nummer: 78-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...