2-Methoxy-N-methylbenzylamine 95%

Leverantör: Apollo Scientific

Synonyms: 1-(2-Methoxyphenyl)-N-methylmethanamine

OR925436-10G OR925436-1G OR925436-5G
APOSOR925436-10GEA 3050 SEK
APOSOR925436-10G APOSOR925436-1G APOSOR925436-5G
2-Methoxy-N-methylbenzylamine 95%
2-Methoxy-N-methylbenzylamine
Formel: C₉H₁₃NO
MW: 151,209 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00025755
CAS nummer: 6851-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR