4-(3-Phenylpropyl)pyridine 95%

Leverantör: Apollo Scientific

OR925286-100G OR925286-25G OR925286-500G
APOSOR925286-100GEA 2150 SEK
APOSOR925286-100G APOSOR925286-25G APOSOR925286-500G
4-(3-Phenylpropyl)pyridine 95%
4-(3-Phenylpropyl)pyridine
Formel: C₁₄H₁₅N
MW: 197,28 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00047458
CAS nummer: 2057-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR