3,3-Dimethyl-1-indanone 95%
Leverantör: Apollo Scientific
Synonyms: 3,3-Dimethyl-2,3-dihydro-1H-inden-1-one
OR925211-10G OR925211-1G OR925211-5G
APOSOR925211-10GEA 1300 SEK
APOSOR925211-10G APOSOR925211-1G APOSOR925211-5G
3,3-Dimethyl-1-indanone 95%
3,3-Dimethyl-1-indanone
Formel: C₁₁H₁₂O
MW: 160,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01846169
CAS nummer: 26465-81-6