Trimethyl orthobenzoate 97%
Leverantör: Apollo Scientific
Synonyms: Trimethyl orthobeznoate, (Trimethoxymethyl)benzene
OR925124-100G OR925124-25G OR925124-500G
APOSOR925124-100GEA 3450 SEK
APOSOR925124-100G APOSOR925124-25G APOSOR925124-500G
Trimethyl orthobenzoate 97%
Trimethyl orthobenzoate
Formel: C₁₀H₁₄O₃
MW: 182,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00008474
CAS nummer: 707-07-3