2-Ethylphenyl isocyanate 95%
Leverantör: Apollo Scientific
OR925112-1G OR925112-5G
APOSOR925112-1GEA 1375 SEK
APOSOR925112-1G APOSOR925112-5G
2-Ethylphenyl isocyanate 95%
2-Ethylphenyl isocyanate
  • May form small amount of sediment.
Formel: C₉H₉NO
MW: 147.18 g/mol
Kokpunkt: 56
Storage Temperature: Omgivande
MDL Number: MFCD00002013
CAS nummer: 40411-25-4