Methyltrimethoxysilane 98%

Leverantör: Apollo Scientific

Synonyms: Trimethoxy(methyl)silane

OR924969-100G OR924969-1KG OR924969-500G
APOSOR924969-100GEA 1475 SEK
APOSOR924969-100G APOSOR924969-1KG APOSOR924969-500G
Methyltrimethoxysilane 98%
Methyltrimethoxysilane
Formel: C₄H₁₂O₃Si
MW: 136,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00008342
CAS nummer: 1185-55-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR