2-Cyclohexylcyclohexanone 95%

Leverantör: Apollo Scientific

OR924905-1G OR924905-25G OR924905-5G
APOSOR924905-1GEA 1375 SEK
APOSOR924905-1G APOSOR924905-25G APOSOR924905-5G
2-Cyclohexylcyclohexanone 95%
2-Cyclohexylcyclohexanone
Formel: C₁₂H₂₀O
MW: 180.29 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00013795
CAS nummer: 90-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR