2-Phenylcyclohexanone 95%
Leverantör: Apollo Scientific
OR924883-1G
OR924883-5G
APOSOR924883-1GEA
855
SEK
APOSOR924883-1G
APOSOR924883-5G
2-Phenylcyclohexanone 95%
2-Phenylcyclohexanone
Formel:
C₁₂H₁₄O MW: 174,24 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00001629 CAS nummer: 1444-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...