1-Pyrrolidinecarbonyl chloride 95%

Leverantör: Apollo Scientific

OR924831-1G OR924831-5G
APOSOR924831-1GEA 255 SEK
APOSOR924831-1G APOSOR924831-5G
1-Pyrrolidinecarbonyl chloride 95%
1-Pyrrolidinecarbonyl chloride
Formel: C₅H₈ClNO
Storage Temperature: Omgivande
MDL Number: MFCD00051321
CAS nummer: 1192-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR