Chloromethyl 2-methylpropanoate 98%

Leverantör: Apollo Scientific
Fara

OR923754-100G OR923754-25G
APOSOR923754-100GEA 2850 SEK
APOSOR923754-100G APOSOR923754-25G
Chloromethyl 2-methylpropanoate 98%
Chloromethyl 2-methylpropanoate
Formel: C₅H₉ClO₂
Storage Temperature: Omgivande
MDL Number: MFCD16038293
CAS nummer: 61644-18-6
UN: 2924
ADR: 3+8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR