3-Methoxyphenol 95%
Leverantör: Apollo Scientific
Synonyms:
3-Metoxifenol
OR923568-100G
OR923568-25G
APOSOR923568-100GEA
1425
SEK
APOSOR923568-100G
APOSOR923568-25G
3-Methoxyphenol 95%
3-Methoxyphenol
Formel:
C₇H₈O₂ MW: 124,14 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00002267 CAS nummer: 150-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...