Dimethyl-trans-1,2-cyclopropanedicarboxylate 95%
Leverantör: Apollo Scientific
Synonyms: trans-cyclopropane-1,2-dicarboxylic acid dimethylester, 1,2-dimethyl (1R,2R)-cyclopropane-1,2-dicarboxylate, (1R,2R)-rel-Dimethyl cyclopropane-1,2-dicarboxylate
OR923179-10G OR923179-1G OR923179-5G
APOSOR923179-10GEA 9500 SEK
APOSOR923179-10G APOSOR923179-1G APOSOR923179-5G
Dimethyl-trans-1,2-cyclopropanedicarboxylate 95%
Dimethyl-trans-1,2-cyclopropanedicarboxylate
Formel: C₇H₁₀O₄
MW: 158,153 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00062806
CAS nummer: 826-35-7